Learn More
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4 (0.1 mg/mL in Methanol), TRC
Product Code. 30388781
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388781 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30388781 Supplier Toronto Research Chemicals Supplier No. TRCKIT07805X1ML
 Restricted Item
Estimated Shipment: 14-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1020719-69-0
Molecular Formula C10 2H4 H7 N3 O
Quantity 5 x 1 mL
Synonym 2,3'-Bipyridine-2',4',5',6'-d4, 1,2,3,6-tetrahydro-1-nitroso-, (RS)-N-Nitrosoanatabine D4 (pyridinyl D4), (R,S)-N-Nitroso Anatabine-2,4,5,6-d4, (R,S)-N-Nitrosoanatabine D4 (pyridinyl D4), 1,2,3,6-Tetrahydro-1-nitroso-2,3'-bipyridine-2',4',5',6'-d4 (ACI)
InChI Formula InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CC=CCN2N=O)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-(1-nitroso-3,6-dihydro-2H-pyridin-2-yl)pyridine
Molecular Weight (g/mol) 193.24
Formula Weight 193.1153
Concentration or Composition (by Analyte or Components) (R,S)-N-Nitroso Anatabine-2,4,5,6-d4 0.1 mg/mL
Chemical Name or Material (R,S)-N-Nitroso Anatabine-2,4,5,6-d4