Learn More
(R,S)-N-Nitroso Anabasine, TRC
Product Code. 30005930
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005930 100 mg 100mg
30009847 50 mg 50mg
18033620 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005930 Supplier Toronto Research Chemicals Supplier No. TRCN524250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37620-20-5
Molecular Formula C10 H13 N3 O
Molecular Weight (g/mol) 191.23
Synonym Pyridine, 3-(1-nitroso-2-piperidinyl)-, Piperidine, 1-nitroso-2-(3-pyridyl)- (6CI,7CI), 3-(1-Nitroso-2-piperidinyl)pyridine, (R,S)-N-Nitroso Anabasine, (RS)-N-Nitrosoanabasine, NAB
IUPAC Name 3-(1-nitrosopiperidin-2-yl)pyridine
SMILES O=NN1CCCCC1c2cccnc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2
Formula Weight 191.1059
Percent Purity >95%
Chemical Name or Material (RS)-N-Nitrosoanabasine
Grade HPLC