Learn More
(R/S)-N-Deacetyl Colchiceine N-Trifluroracetate, TRC
Product Code. 30359394
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359394 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30359394 Supplier Toronto Research Chemicals Supplier No. TRCD19890550MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R/S)-N-Deacetyl Colchiceine N-Trifluroracetate
CAS 102491-70-3
Molecular Formula C21H20F3NO6
Quantity 50 mg
Synonym 2,2,2-Trifluoro-N-[5,6,7,9-tetrahydro-10-hydroxy-1,2,3-trimethoxy-9-oxobenzo[a]heptalen-7-yl]-acetamide
InChI Formula InChI=1S/C21H20F3NO6/c1-29-16-8-10-4-6-13(25-20(28)21(22,23)24)12-9-15(27)14(26)7-5-11(12)17(10)19(31-3)18(16)30-2/h5,7-9,13H,4,6H2,1-3H3,(H,25,28)(H,26,27)
SMILES COc1cc2CCC(NC(=O)C(F)(F)F)C3=CC(=O)C(=CC=C3c2c(OC)c1OC)O
IUPAC Name 2,2,2-trifluoro-N-(10-hydroxy-1,2,3-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl)acetamide
Molecular Weight (g/mol) 439.38
Formula Weight 439.1243