Learn More
R&D Systems™ Quantikine Immunoassay Control Set 851 Human Free IGFBP-1
Product Code. 30001971
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001971 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001971 Supplier R&D Systems™ Supplier No. QC204
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / Pack of 1
    Save 23.48€
    10% Off
 Restricted Item
Estimated Shipment: 17-11-2026
For use with catalog number DGB100
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes IGFBP-1
Gene ID (Entrez) 3484
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human