Learn More
R&D Systems™ Quantikine Immunoassay Control Set 786 for Human Cyr61/CCN1
Product Code. 18481990
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18481990 1 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18481990 Supplier R&D Systems™ Supplier No. QC73
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / 1 sets
    Save 23.48€
    10% Off
 Restricted Item
Estimated Shipment: 23-11-2026
For use with catalog number DCYR10
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Cyr61 (CCN1)
Gene ID (Entrez) 3491
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human