Learn More
R&D Systems™ Quantikine Immunoassay Control Set 717 for Human ADAMTS13
Product Code. 18742338
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18742338 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18742338 Supplier R&D Systems™ Supplier No. QC35
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / Pack of 1
    Save 23.48€
    10% Off
 Restricted Item
Estimated Shipment: 02-12-2026
For use with catalog number DADT130
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes ADAMTS13
Gene ID (Entrez) 11093
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human