missing translation for 'onlineSavingsMsg'
Learn More

(R)-Oxybutynin Chloride, TRC

Product Code. 30423985
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423985 10 mg 10mg
30424651 50 mg -
30415916 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423985 Supplier Toronto Research Chemicals Supplier No. TRCO86851010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207344-05-5
Molecular Formula C22 H31 N O3 . Cl H
Molecular Weight (g/mol) 393.95
Synonym Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(diethylamino)-2-butyn-1-yl ester, hydrochloride (1:1), (αR)- (ACI), 4-(Diethylamino)-2-butyn-1-yl (αR)-α-cyclohexyl-α-hydroxybenzeneacetate hydrochloride (1:1) (ACI), (R)-Oxybutynin hydrochloride, 4-(Diethylamino)-2-butynyl (αR)-α-phenylcyclohexaneglycolate hydrochloride, Aroxybutynin hydrochloride
IUPAC Name 4-(diethylamino)but-2-ynyl (2R)-2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride
SMILES Cl.CCN(CC)CC#CCOC(=O)[C@@](O)(C1CCCCC1)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H/t22-;/m0./s1
Formula Weight 393.2071
Chemical Name or Material (R)-Oxybutynin Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.