missing translation for 'onlineSavingsMsg'
Learn More

(R)-Naproxen Acyl-Beta-D-glucuronide, TRC

Product Code. 30473345
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473345 1 mg 1mg
30481806 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473345 Supplier Toronto Research Chemicals Supplier No. TRCN3775151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112828-15-6
Molecular Formula C20 H22 O9
Molecular Weight (g/mol) 406.38
Synonym (R)-Naproxen Acyl-Ss-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(2R)-2-(6-methoxynaphthalen-2-yl)propanoyl]oxyoxane-2-carboxylic acid
SMILES COc1ccc2cc(ccc2c1)[C@@H](C)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H22O9/c1-9(10-3-4-12-8-13(27-2)6-5-11(12)7-10)19(26)29-20-16(23)14(21)15(22)17(28-20)18(24)25/h3-9,14-17,20-23H,1-2H3,(H,24,25)/t9-,14+,15+,16-,17+,20+/m1/s1
Formula Weight 406.1264
Chemical Name or Material (R)-Naproxen Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.