Learn More
(R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate, 95%, Thermo Scientific™
Product Code. 11461130
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11461130
Brand: Thermo Scientific Alfa Aesar H25778.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C18H25FeNO6
MDL Number MFCD09953453
Specifications
Melting Point 129°C to 131°C
Quantity 25 g
Optical Rotation +14° (c=1 in Methanol)
Formula Weight 407.26
Percent Purity 95%
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate