Learn More
(R)-N-Benzyl-3-methoxy-2-(N-methylacetamido)propanamide, TRC
Product Code. 30133633
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133633 100 mg 100mg
30134598 10 mg 10mg
30006200 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133633 Supplier Toronto Research Chemicals Supplier No. TRCM315215100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2742962-60-1
Molecular Formula C14H20N2O3
Molecular Weight (g/mol) 264.32
Synonym (R)-N-Benzyl-2-[N-acetyl N-Methyl-amino]-3-methoxypropionamide;
IUPAC Name (2R)-2-[acetyl(methyl)amino]-N-benzyl-3-methoxypropanamide
SMILES O=C([C@H](N(C(C)=O)C)COC)NCC1=CC=CC=C1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H20N2O3/c1-11(17)16(2)13(10-19-3)14(18)15-9-12-7-5-4-6-8-12/h4-8,13H,9-10H2,1-3H3,(H,15,18)/t13-/m1/s1
Formula Weight 264.15
Percent Purity >95%
Chemical Name or Material (2R)-N-Benzyl-3-methoxy-2-(N-methylacetamido)propanamide
Grade HPLC