missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Neuropilin-1 Quantikine ELISA Kit

Product Code. 18761844
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18761844 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18761844 Supplier R&D Systems™ Supplier No. DNRP10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Neuropilin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Neuropilin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Neuropilin-1 in cell culture supernates (50 ul), cell lysates (13 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul), human milk (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 9.33pg/mL
Detectable Analytes Neuropilin-1
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human Neuropilin-1
For Use With (Application) ELISA
Gene ID (Entrez) 8829
Host Species Human
Measuring Range 78.1-5,000pg/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (13uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.