missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Iduronate 2-Sulfatase/IDS DuoSet ELISA

Product Code. 30021272
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021272 1 Kit (for 5 Plates) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021272 Supplier R&D Systems™ Supplier No. DY244905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Human Iduronate 2-Sulfatase/IDS DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Iduronate 2-Sulfatase/IDS.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.10 to 5000 pg/mL
Detectable Analytes Iduronate 2-Sulfatase/IDS
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 3423
Gene Symbol IDS
Host Species Human
Measuring Range 78.1-5,000pg/mL
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS
Target Iduronate 2-Sulfatase/IDS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.