missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TACS Annexin V-FITC Apoptosis Detection Kit

Product Code. p-4830549
Click to view available options
Quantity:
100 Tests
250 Tests
Unit Size:
25 tests
250 tests
This item is not returnable. View return policy

Product Code. 18421830

Brand: R&D Systems™ 483001K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

TACS Annexin V-FITC, 250uL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL

Facilitates rapid fluorometric quantitation of apoptotic cells

Early in the apoptosis process, phosphatidylserine (PS) becomes exposed on the cell surface by flipping from the inner to outer leaflet of the cytoplasmic membrane. This event is thought to be important for macrophage recognition of cells undergoing apopt

Supplied with an optimized binding buffer and propidium iodide, included in the incubation mix to identify cells that have lost membrane integrity
As cells disintegrate, greater access to the inner cell membrane allows for additional Annexin V binding: T

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Kit Contents TACS Annexin V-FITC, 100μL; 10X Binding Buffer, 8mL; Propidium Iodide, 1mL
Product Type TACS Annexin V-FITC Apoptosis Detection Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Facilitates rapid fluorometric quantitation of apoptotic cells
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.