Learn More
R&D Systems™ StemXVivo Serum-Free Human MSC Expansion Media
Product Code. 18431640
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431640 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18431640 Supplier R&D Systems™ Supplier No. CCM014
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
A complete media formulated and optimized for the maintenance and expansion of purified human MSCs
Mesenchymal stem/stromal cells (MSCs) are a rare population of multipotent cells that can be derived from a number of tissues such as bone marrow, adipose, and placenta.
  • Lacks serum to reduce experimental variation
  • Is optimized to support reproducible human MSC growth in vitro
  • Can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
  • Is suitable for transitioning cultures to pre-clinical expansion protocols
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Expansion Media
Quantity 500 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.