Learn More
R&D Systems™ StemXVivo Methylcellulose Concentrate
Product Code. 18472630
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18472630 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18472630 Supplier R&D Systems™ Supplier No. HSC011
 Restricted Item
Estimated Shipment: 06-11-2026
For the growth and differentiation of human, mouse and rat hematopoietic stem cells using Iscove'ss Modified Dulbecco's Media or any other basal media
TRUSTED_SUSTAINABILITY
Specifications
Product Type Methylcellulose Concentrate, semisolid medium
Product Line R&D Systems
Quantity 50 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.