Learn More
R&D Systems™ StemXVivo Mesenchymal Stem Cell Expansion Media
Product Code. 18481960
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18481960 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18481960 Supplier R&D Systems™ Supplier No. CCM004
  • ONLINE EXCLUSIVE PRICE
    256.54€
    228.90€ / 250mL
    Save 27.64€
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
A complete, fetal bovine serum-containing media formulated and optimized for the maintenance and expansion of purified human, mouse and rat MSCs
  • Supports reproducible MSC growth in vitro with a formula to reduce variation
  • Ready-to-use formula can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
  • Has been developed and optimized for use with human, mouse, and rat MSCs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Expansion Media
Product Line R&D Systems
Quantity 250 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.