Learn More
R&D Systems™ Proteome Profiler Rat Cytokine Array Kit, Panel A
GREENER_CHOICE
Product Code. 18087641
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18087641 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18087641 Supplier R&D Systems™ Supplier No. ARY008
  • ONLINE EXCLUSIVE PRICE
    684.00€
    646.80€ / 1 sets
    Save 37.20€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
The Proteome Profiler Rat Cytokine Array Kit, Panel A requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Rat Cytokine Array Kit, Panel A contains 4 membranes - each spotted in duplicate with 29 different cytokine antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit