Learn More
R&D Systems™ Proteome Profiler Mouse Angiogenesis Array Kit
Product Code. 18087631
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18087631
Brand: R&D Systems™ ARY015
The Proteome Profiler Mouse Angiogenesis Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Mouse Angiogenesis Array Kit contains 4 membranes - each spotted in duplicate with 53 different angiogenesis antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit