Learn More
R&D Systems™ Proteome Profiler Human Phospho-Immunoreceptor Array Kit
GREENER_CHOICE
Product Code. 18007621
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18007621 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18007621 Supplier R&D Systems™ Supplier No. ARY004B
  • ONLINE EXCLUSIVE PRICE
    732.00€
    691.95€ / 1 sets
    Save 40.05€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
The Proteome Profiler Human Phospho-Immunoreceptor Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Phospho-Immunoreceptor Array Kit contains 4 membranes - each spotted in duplicate with 59 different immunoreceptor antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit