Learn More
R&D Systems™ Proteome Profiler Human Angiogenesis Array Kit
GREENER_CHOICE
Product Code. 18097611
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18097611 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18097611 Supplier R&D Systems™ Supplier No. ARY007
  • ONLINE EXCLUSIVE PRICE
    844.00€
    798.00€ / 1 sets
    Save 46.00€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
The Proteome Profiler Human Angiogenesis Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Angiogenesis Array Kit contains 4 membranes - each spotted in duplicate with 55 different angiogenesis antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit