Learn More
R&D Systems™ pERTKR-AMC Fluorogenic Peptide Substrate
Product Code. 18377497
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377497 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18377497 Supplier R&D Systems™ Supplier No. ES013
  • ONLINE EXCLUSIVE PRICE
    316.00€
    298.20€ / 10mg
    Save 17.80€
    5% Off
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
Substrate for Furin, Proprotein Convertase 1 and 7, and wnvNS3
  • Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Purity >96%
  • Excellent substrate for furin-like proprotein convertases
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type L-PyroGlu-Arg-Thr-Lys-Arg-AMC
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.