Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Product Code. p-4830531
Click to view available options
Quantity:
2500 Tests
5000 Tests
Unit Size:
2500 tests
5000 tests
Product Code. 18442630
Brand: R&D Systems™ 4890050K
Includes
MTT reagent; Detergent Reagent
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5000 Tests
Product Type MTT Cell Proliferation/Viability Assay
Content And Storage The product requires storage at 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
For Use With (Application) Minimizes steps to assay for proliferation rate in cultured cells