Learn More
R&D Systems™ MTT Cell Proliferation/Viability Assay
Description
Includes
MTT reagent; Detergent Reagent
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
Specifications
| Quantity | 2500 Tests |
| Product Type | MTT Cell Proliferation/Viability Assay |
| Content And Storage | The product requires storage at 2°C to 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| For Use With (Application) | Minimizes steps to assay for proliferation rate in cultured cells |