Learn More
R&D Systems™ Human MCT8/SLC16A2 (NP_006508) VersaClone cDNA
Product Code. 18732559
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732559 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18732559 Supplier R&D Systems™ Supplier No. RDC0892
  • ONLINE EXCLUSIVE PRICE
    362.41€
    322.35€ / 10µg
    Save 40.06€
    11% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Monocarboxylic Acid Transporter 8 (Solute Carrier Family 16 Member 1)
The Human MCT8/SLC16A2 (NP_006508) VersaClone cDNA clone contains the complete ORF for MCT8/SLC16A2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MCT8/SLC16A2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Gene hSLC16A2
Accession Number NP_006508
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1632bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans