missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD1e (CAB93150) VersaClone cDNA

Product Code. 18771319
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18771319 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18771319 Supplier R&D Systems™ Supplier No. RDC1273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HM31; ILT-6; ILT6CD85e; Immunoglobulin-like transcript 6; leucocyte immunoglobulin-like receptor

The Human CD1e (CAB93150) VersaClone cDNA clone contains the complete ORF for ILT6/CD85e/LILRA3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ILT6/CD85e/LILRA3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA
Gene hCD1E
Accession Number CAB93150
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1180bp
Gene ID (Entrez) 11026
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.