Learn More
R&D Systems™ Cynomolgus Monkey PD-L2/B7-DC (XP_005581839) VersaClone cDNA
Description
The Cynomolgus Monkey PD-L2/B7-DC (XP_005581839) VersaClone cDNA clone contains the complete ORF for PD-L2/B7-DC/PDCD1LG2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PD-L2/B7-DC/PDCD1LG2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Product Type | cDNA |
| Description | Gene ID (Entrez): 80380, 58205, 102146227 |
| Gene | PD-L2/B7-DC/PDCD1LG2 |
| Accession Number | XP_005581839 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 835bp |
| Gene ID (Entrez) | 80380 |
| Gene Alias | B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Show More |
For Research Use Only; Not for Use in Humans