Learn More
R&D Systems™ Cultrex 3-D Culture Matrix RGF Basement Membrane Extract
Product Code. 18481270
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18481270 1 mL 1mL
18462400 2 x 5 mL 10mL
18462020 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18481270 Supplier R&D Systems™ Supplier No. 344500101
  • ONLINE EXCLUSIVE PRICE
    115.04€
    101.85€ / 1mL
    Save 13.19€
    11% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Applications: Bioactivity
Functional assays: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. 3D Culture - Basement Membrane Extract promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) or human prostate (PC-3) into acinar structures. Gelling assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 8-12 mg/mL (by Lowry assay)
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 1 mL
Endotoxin Level ≤8EU/mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.