Learn More
R&D Systems™ Cultrex 3-D Culture Matrix RGF Basement Membrane Extract
Product Code. 18462400
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Unit Size:
10mL
1mL
5mL
Product Code. 18462400
Brand: R&D Systems™ 344501001
Applications: Bioactivity
Functional assays: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. 3D Culture - Basement Membrane Extract promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) or human prostate (PC-3) into acinar structures. Gelling assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 14 to 16 mg/mL
For Use With (Application) Bioactivity
Quantity 2 x 5 mL
Endotoxin Level ≤8EU/mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.