Learn More
R&D Systems™ Ac-Arg-Gly-Lys(Ac)-AMC Fluorogenic Peptide Substrate
Product Code. 18108320
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18108320 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18108320 Supplier R&D Systems™ Supplier No. ES016
  • ONLINE EXCLUSIVE PRICE
    316.00€
    298.20€ / 5mg
    Save 17.80€
    5% Off
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
Substrate for certain deacetylases such as histone deacetylases
  • Hydrolysis of Lys-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • After the enzymatic removal of the acetyl group, the substrate can be cleaved by trypsin-like enzymes
  • Purity >95%
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mg
Product Type Ac-Arg-Gly-Lys(Ac)-AMC (Ac-RGK(Ac)-AMC)
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.