missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R)-CR8
GREENER_CHOICE

Product Code. 18034532
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18034532 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18034532 Supplier Tocris Bioscience™ Supplier No. 3605/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk1 and cdk2; also inhibits CK1; acts as molecular glue

(R)-CR8 is a cyclin dependent kinase (cdk) inhibitor (reported IC50 values are 0.036 - 0.07, 0.09 - 0.8, 0.13 - 0.68, 0.18 - 1 and >1 μM for cdk2, cdk1, cdk5, cdk9 and cdk7, respectively). Also inhibits casein kinase 1 (CK1; IC50 = 0.6 μM). Acts as molecular glue; forms a complex between CDK12-cyclin K and the CUL4 adapter protein DDB1 resulting in ubiquination and degradation of cyclin K.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1786438-30-9
Quantity 10 mg
Molecular Formula C24H29N7O0.3 HCl
Product Type (2R)-2-[[9-(Methylethyl)-6-[[[4-(2-pyridinyl)phenyl]methyl]amino]-9H-purin-2-yl]amino]-1-butanol trihydrochloride
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 540.92
Inhibitors (R)-CR8

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.