Learn More
(R)-(+)-Citronellic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-(+)-Citronellic Acid |
| CAS | 18951-85-4 |
| Molecular Formula | C10H18O2 |
| Quantity | 500 mg |
| Synonym | (R)-Citronellic acid, d-Citronellic acid, (+)-(R)-Citronellic acid, (+)-Citronellic acid, (R)-(+)-3,7-Dimethyl-6-octenoic acid, (R)-(+)-3,7-Dimethyl-6-octenoic Acid, (R)-3,7-Dimethyl-6-octenoic Acid, (3R)-3,7-Dimethyl-6-octenoic Acid |
| InChI Formula | InChI=1S/C10H18O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,9H,4,6-7H2,1-3H3,(H,11,12)/t9-/m1/s1 |
| SMILES | OC(CC(C)CC/C=C(C)\C)=O |
| IUPAC Name | (3R)-3,7-dimethyloct-6-enoic acid |
| Molecular Weight (g/mol) | 170.25 |