missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Amlodipine-d4 Di-O-benzoyl L-Tartaric Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Amlodipine-d4 Di-O-benzoyl L-Tartaric Acid Salt |
| Molecular Formula | C20 D4 H21 Cl N2 O5 . C18 H14 O8 |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C20H25ClN2O5.C18H14O8/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12/h5-8,17,23H,4,9-11,22H2,1-3H3;1-10,13-14H,(H,19,20)(H,21,22)/t17-;13-,14-/m11/s1/i9D2,10D2; |
| SMILES | [2H]C([2H])(N)C([2H])([2H])OCC1=C([C@@H](C(=C(C)N1)C(=O)OC)c2ccccc2Cl)C(=O)OCC.OC(=O)[C@H](OC(=O)c3ccccc3)[C@@H](OC(=O)c4ccccc4)C(=O)O |
| IUPAC Name | (2R,3R)-2,3-dibenzoyloxybutanedioic acid;3-O-ethyl 5-O-methyl (4R)-2-[(2-amino-1,1,2,2-tetradeuterioethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Molecular Weight (g/mol) | 771.199 |
| Formula Weight | 770.239 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?