missing translation for 'onlineSavingsMsg'
Learn More

R 428, TRC

Product Code. 30410647
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410647 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410647 Supplier Toronto Research Chemicals Supplier No. TRCR0600201MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(6,7-Dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazin-3-yl)-N3-((7-(S)-pyrrolidin-1-yl)-6,7,8,9-tetrahydro-5H-benzo[7]annulene-2-yl)-1H-1,2,4-triazole-3,5-diamine
CAS 1037624-75-1
Molecular Formula C30H34N8
Quantity 1 mg
Synonym BGB 324, R 428
SMILES Nc1nc(Nc2ccc3CC[C@@H](CCc3c2)N4CCCC4)nn1c5cc6CCCc7ccccc7c6nn5
IUPAC Name 1-(6,7-dihydro-5H-benzo[4,5]cyclohepta[1,2-d]pyridazin-3-yl)-3-N-[(7S)-7-pyrrolidin-1-yl-6,7,8,9-tetrahydro-5H-benzo[7]annulen-3-yl]-1,2,4-triazole-3,5-diamine
Molecular Weight (g/mol) 506.64
Formula Weight 506.2906
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.