Learn More
(R)-(-)-4-Penten-2-ol, 95%
Product Code. 11339919
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339919 1 g 1g
11349919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11339919 Supplier Thermo Scientific Alfa Aesar Supplier No. B25659.03
Request Bulk or Custom Format
CAS: 64584-92-5 | C5H10O | 86.13 g/mol
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
Solubility
Soluble in water, 4.526 mg/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64584-92-5
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD03701536
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYNA-N
Synonym r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
PubChem CID 6999724
SMILES CC(O)CC=C
Specifications
Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.424
Quantity 1 g
UN Number UN1987
Solubility Information Soluble in water,4.526mg/L @ 25°C.
Optical Rotation −5.2° (c=1 in Chloroform)
IUPAC Name pent-4-en-2-ol
Formula Weight 86.13
Percent Purity 95%
Chemical Name or Material (R)-(-)-4-Penten-2-ol