missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-Benzyl-2-oxazolidinone, 99%

Product Code. 11462788
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11462788

Brand: Thermo Scientific Alfa Aesar A16770.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102029-44-7 | C10H11NO2 | 177.20 g/mol

(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-4-Benzyl-2-oxazolidinones are used in the enantioselective synthesis of beta-lactams and alpha-amino acids. End products can include asysmmetric single isomeric drugs of antitumor, anesthetic, antipasmodic products. Used in the synthesis of HIV protease inhibitors. An oxazolidinone used in chiral auxiliary asymetric synthesis.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102029-44-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00010846
InChI Key OJOFMLDBXPDXLQ-SECBINFHSA-N
Synonym r-4-benzyl-2-oxazolidinone, r-+-4-benzyl-2-oxazolidinone, r-4-benzyloxazolidin-2-one, 4r-4-benzyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4r, r---4-benzyl-2-oxazolidinone, r-4-benzyl-oxazolidin-2-one, 4-benzyl-1,3-oxazolidin-2-one #
PubChem CID 2734969
IUPAC Name (4R)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 86°C to 89°C
Beilstein 4782551
Solubility Information Insoluble in water. Soluble in Chloroform.
Quantity 5 g
Formula Weight 177.2
Optical Rotation +64° (c=1 in Chloroform)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material (R)-(+)-4-Benzyl-2-oxazolidinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.