Learn More
(R)-(+)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-oxazolidinecarboxylic Acid Methyl Ester, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (R)-(+)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-oxazolidinecarboxylic Acid Methyl Ester |
| CAS | 95715-86-9 |
| Molecular Formula | C12H21NO5 |
| Quantity | 1 g |
| Synonym | (4R)-2,2-Dimethyl-3,4-oxazolidinedicarboxylic Acid 3-(1,1-Dimethylethyl) 4-Methyl Ester, (R)-2,2-Dimethyloxazolidine-3,4-dicarboxylic Acid 3-tert-Butyl Ester 4-Methyl Ester |
| InChI Formula | InChI=1S/C12H21NO5/c1-11(2,3)18-10(15)13-8(9(14)16-6)7-17-12(13,4)5/h8H,7H2,1-6H3/t8-/m1/s1 |
| SMILES | COC(=O)[C@H]1COC(C)(C)N1C(=O)OC(C)(C)C |
| IUPAC Name | 3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate |
| Molecular Weight (g/mol) | 259.3 |
| Formula Weight | 259.142 |