missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Aminobutanoic Acid, TRC

Product Code. 30309403
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309403 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309403 Supplier Toronto Research Chemicals Supplier No. TRCA60294810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-3-Aminobutanoic Acid
CAS 3775-73-3
Molecular Formula C4H9NO2
Quantity 10 g
Synonym (3R)-3-Aminobutanoic Acid, (-)-3-Aminobutanoic Acid, (-)-3-Aminobutyric Acid, (3R)-3-Azaniumylbutanoate, (R)-(-)-3-Aminobutyric Acid, (R)-3-Aminobutanoic Acid, (R)-3-Aminobutanoic Acid, (R)-3-Aminobutyric Acid, (R)-beta-Aminobutanoic Acid, 3(R)-Methyl-beta-alanine, D-3-Aminobutanoic Acid, D-3-Aminobutyric Acid, D-beta-Aminobutyric Acid
InChI Formula InChI=1S/C4H9NO2/c1-3(5)2-4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m1/s1
SMILES C[C@@H](N)CC(=O)O
IUPAC Name (3R)-3-aminobutanoic acid
Molecular Weight (g/mol) 103.11
Formula Weight 103.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.