missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-Tosyloxy-2,3-propanediol-d2, TRC
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Specifications
Specifications
| Chemical Name or Material | (R)-1-Tosyloxy-2,3-propanediol-d2 |
| Molecular Formula | C10 D2 H12 O5 S |
| Quantity | 5 mg |
| Synonym | (R)-3-(Tosyloxy)-1,2-propanediol-d2, (R)-3-(p-Toluenesulfonyloxy)-1,2-propanediol-d2, (R)-1,2,3-Propanetriol 4-methylbenzenesulfonate-d2, (R)-1-Tosyloxyglycerol-d2 |
| InChI Formula | InChI=1S/C10H14O5S/c1-8-2-4-10(5-3-8)16(13,14)15-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3/t9-/m0/s1/i7D2 |
| SMILES | [2H]C([2H])(OS(=O)(=O)c1ccc(C)cc1)[C@@H](O)CO |
| IUPAC Name | [(2S)-1,1-dideuterio-2,3-dihydroxypropyl] 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 248.292 |
| Formula Weight | 248.069 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction