missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-(4-Chlorophenyl)ethyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 15420867
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420867 Supplier Thermo Scientific Alfa Aesar Supplier No. L19974.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737000-80-5
Density 1.208
Quantity 1 g
Refractive Index 1.594
Molecular Formula C9H8ClNS
MDL Number MFCD04039326
UN Number UN2922
Synonym r---1-4-chlorophenyl ethyl isothiocyanate, 1-chloro-4-1r-1-isothiocyanatoethyl benzene
InChI Key BQJKNZREKDQMQA-UHFFFAOYNA-N
SMILES CC(N=C=S)C1=CC=C(Cl)C=C1
IUPAC Name 1-chloro-4-[(1R)-1-isothiocyanatoethyl]benzene
Molecular Weight (g/mol) 197.68
PubChem CID 7016319
Formula Weight 197.69
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-(4-Chlorophenyl)ethyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.