missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. 11382749
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11382749

Brand: Thermo Scientific Alfa Aesar H27673.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23357-47-3 | C11H12O2 | 176.215 g/mol

It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23357-47-3
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368364
InChI Key VDLWTJCSPSUGOA-SNVBAGLBSA-N
Synonym r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid
PubChem CID 12598147
IUPAC Name (1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Optical Rotation +12° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (R)-1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.