missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogen Phosphate, TRC

Product Code. 30330746
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330746 Supplier Toronto Research Chemicals Supplier No. TRCB3870001G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogen Phosphate
CAS 39648-67-4
Molecular Formula C20 H13 O4 P
Quantity 1 g
Synonym Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 4-hydroxy-, 4-oxide, (11bR)-, Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 4-hydroxy-, 4-oxide, (R)-, (-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, (-)-1,1'-Dinaphthyl-2,2'-diyl hydrogen phosphate, (-)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide, (R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, (R)-(-)-BNDHP, (R)-(-)-Binaphthyl-2,2'-diyl hydrogen phosphate, (R)-1,1'-Bi-2-naphthol-2,2'-diyl hydrogen phosphate, (R)-1,1'-Binaphthalene-2,2'-diyl hydrogen phosphate, (R)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, (R)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, (R)-BNPPA
InChI Formula InChI=1S/C20H13O4P/c21-25(22)23-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)24-25/h1-12H,(H,21,22)
SMILES OP1(=O)Oc2ccc3ccccc3c2c4c(O1)ccc5ccccc45
Molecular Weight (g/mol) 348.29
Formula Weight 348.0551
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.