Learn More
QX 572, TRC
Specifications
Specifications
| Chemical Name or Material | QX 572 |
| CAS | 1042-42-8 |
| Molecular Formula | C18 H22 N3 O2 . Cl |
| Quantity | 1 g |
| Synonym | N,N-Dimethyl-2-oxo-N-[2-oxo-2-(phenylamino)ethyl]-2-(phenylamino)-ethanaminium Chloride, Dimethylbis[(phenylcarbamoyl)methyl]-ammonium Chloride, Dimethylbis[(phenylcarbamoyl)methyl]ammonium Chloride, Carcainium Chloride, N,N-Bis(phenylcarbamoylmethyl)dimethylammonium Chloride, RSD 931 |
| InChI Formula | InChI=1S/C18H21N3O2.ClH/c1-21(2,13-17(22)19-15-9-5-3-6-10-15)14-18(23)20-16-11-7-4-8-12-16;/h3-12H,13-14H2,1-2H3,(H-,19,20,22,23);1H |
| SMILES | [Cl-].C[N+](C)(CC(=O)Nc1ccccc1)CC(=O)Nc2ccccc2 |
| IUPAC Name | bis(2-anilino-2-oxoethyl)-dimethylazanium;chloride |
| Molecular Weight (g/mol) | 347.84 |
| Formula Weight | 347.1401 |