Learn More
Quickfit™ Straight Delivery Adapter
Description
Deliver liquids securely with glass straight receivers. QuickFit™ straight delivery adapter attaches securely to glass containers for transfer.
- Delivery receiver with straight stem
Specifications
Specifications
| Height (English) | 7.08in. |
| Height (Metric) | 180mm |
| Material | Borosilicate Glass |
| Product Type | Straight delivery adapter |
| Standard Taper Joint No. | 19/26 |