Learn More
Quickfit™ Sloping Splash Head Adapter
Product Code. 15587970
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 (Flask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
29/32 (Condenser), 29/32 (Flask)
Unit Size:
Pack of 5
Product Code. 15587970
Brand: Quickfit SH7/03
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Used in distillation assemblies.
  • Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
For Use With (Application) Distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser
Cone Size 19/26 (Condenser), 24/29 (Flask)
Product Type Sloping Splash Head