Learn More
Quickfit™ Reduction Socket Adapter
missing translation for 'orderingAttributeHoverText'
Cone Size:
Cone Size: 14/23
Cone Size: 19/26
Cone Size: 24/29
Cone Size: 29/32
Cone Size: 34/35
Cone Size: 40/38
Cone Size: 45/40
Cone Size: 50/42
Cone Size: 55/44
Standard Taper Joint No.:
10/19
14/23
19/26
24/29
29/32
34/35
missing translation for 'unitSize'
Each
Pack of 2
Pack of 5
Descripción
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Especificaciones
Especificaciones
| Cone Size | Cone Size: 45/40 |
| For Use With (Equipment) | Dissimilar sized ground Glass joint |
| Includes | Small socket, large cone |
| Material | Borosilicate Glass |
| Product Type | Reduction adapter |
| Standard Taper Joint No. | 34/35 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido