Learn More
Quickfit™ Reduction Socket Adapter
Product Code. 12520337
Click to view available options
Cone Size:
Cone Size: 14/23
Cone Size: 19/26
Cone Size: 24/29
Cone Size: 29/32
Cone Size: 34/35
Cone Size: 40/38
Cone Size: 45/40
Cone Size: 50/42
Cone Size: 55/44
Standard Taper Joint No.:
10/19
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 12520337
Brand: Quickfit DA01
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
TRUSTED_SUSTAINABILITY
Specifications
Cone Size Cone Size: 14/23
For Use With (Equipment) Dissimilar sized ground Glass joint
Includes Small socket, large cone
Material Borosilicate Glass
Product Type Reduction adapter
Standard Taper Joint No. 10/19