Learn More
Quickfit™ Distillation Splash Head Adapter
Product Code. 12435396
Change view
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 Fflask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Unit Size
12435396 24/29 (Condenser), 24/29 (Flask) Each
12078493 19/26 (Condenser), 19/26 Fflask) Each
12415396 19/26 (Condenser), 24/29 (Flask) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12435396 Supplier Quickfit Supplier No. SH7/53
  • ONLINE EXCLUSIVE PRICE
    94.29€
    81.52€ / Pack of 1
    Save 12.77€
    13% Off
Estimated Shipment: 30-11-2026
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour stream
Specifications
Material Borosilicate Glass (3.3)
For Use With (Application) Distillation assemblies in which the condenser is assembled in the vertical mode
Cone Size 24/29 (Condenser), 24/29 (Flask)
Product Type Distillation Splash Head Adapter