Learn More
Quickfit™ Distillation Splash Head Adapter
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 Fflask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
Conditionnement:
Each
Description
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour stream
Spécification
Spécification
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Distillation assemblies in which the condenser is assembled in the vertical mode |
| Cone Size | 19/26 (Condenser), 19/26 Fflask) |
| Product Type | Distillation Splash Head Adapter |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu