Learn More
Quickfit™ Chromatography Column with Rotaflo™ Stopcock
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
500 mm
Diameter (Metric) Inner:
10 mm
20 mm
26 mm
40 mm
Connector Types:
14/23 Socket
19/26 Socket
24/29 Socket
40/38 Socket
Includes:
GP Rotaflo 3mm Bore Stopcock
GP Rotaflo 6mm Bore Stopcock
Unit Size:
Each
Description
Manufactured from Pyrex™ borosilicate glass. Quickfit™ Chromatography Column with Rotaflo™ Stopcock includes a 0 porosity chemically resistant glass sintered disc. PTFE Stopcock provides fine control of eluent flow. Ground glass socket accepts Quickfit™ reservoirs.
- With PTFE Rotaflo™ Stopcock for fine control of eluent flow
- Chemically resistant glass sintered disc - porosity 0
- Ground glass socket accepts Quickfit™ reservoirs
- Manufactured from Pyrex™ borosilicate glass
Specifications
Specifications
| Quantity | 1 Pack |
| Diameter (Metric) Inner | 26 mm |
| Type | Chromatography Column |
| Length (Metric) | 500 mm |
| Material | Pyrex™ Borosilicate Glass |
| Connector Types | 24/29 Socket |
| Includes | GP Rotaflo 3mm Bore Stopcock |