Learn More
Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask
Product Code. 10302230
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
700 mL
1000 mL
2000 mL
3000 mL
4 L
6 L
10 L
20 L
Height (Metric):
90 mm
115 mm
135 mm
155 mm
185 mm
210 mm
225 mm
260 mm
320 mm
380 mm
Unit Size:
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Unit Size
10302230 1000 mL 155 mm Each
10696675 250 mL 90 mm Each
11351975 500 mL 115 mm Each
15557900 700 mL 135 mm Pack of 5
11311955 2000 mL 185 mm Each
11351965 3000 mL 210 mm Each
15547890 4 L 225 mm Pack of 5
15547900 6 L 260 mm Pack of 5
11971279 10 L 320 mm Each
11911289 20 L 380 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10302230 Supplier Quickfit Supplier No. FR1LF
  • ONLINE EXCLUSIVE PRICE
    124.95€
    103.45€ / Pack of 1
    Save 21.50€
    17% Off
In Stock
Manufactured from borosilicate glass. Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask's wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.
Manufactured from borosilicate glass
  • With large diameter Pyrex Labware™ Quickfit™ flat-flange joint
  • For use with Pyrex Labware™ Quickfit™ flat-flange lids and retaining clips
  • Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
For Use With (Application) For use with Quickfit™ flat-flange lids and retaining clips
Height (Metric) 155 mm
Neck Style Wide
Bottom Shape Round